accessibility menu, dialog, popup

UserName

CH3OF

CAS RN 1514-26-7

IUPAC Name: 1-(4-fluorophenoxy)-3-methylbenzene
Synonyms: 1-(4-fluorophenoxy)-3-methylbenzene
Molecular Weight (g/mol): 202.23
Molecular Formula: C13H11FO
InChi Key: APVQRVSBMIDSFS-UHFFFAOYSA-N
SMILES: CC1=CC(OC2=CC=C(F)C=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results