missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix Y, Bulk

Lysing Matrix Y is composed of 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads and is used primarily for lysis of gram positive and gram negative bacteria, fungal tissue and spores, yeasts, molds, filamentous algaes and biofilms.

$520.21 - $520.21

Specifications

Bead Type 0.5 mm Yttria-stabilized Zirconium Oxide Beads
Sample Type Yeast, Fungi, Algea
For Use With (Equipment) FastPrep instruments or other bead beaters
For Use With (Application) Nucleic Acid Isolation and Purification, Homogenization
Matrix Lysing Matrix Y
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
116540436
View Documents
MP Biomedicals™
116540436
500 g
Each for $520.21
Only null left
 
Description

Description

  • Applications: Lysing Matrix Y is used primarily for lysis of gram positive and gram negative bacteria, fungal tissue and spores, yeasts, molds, filamentous algaes and biofilms.
  • The beads have a high density and an extremely high hardness making this matrix ideal for cracking of very tough cell walls while its smooth surface minimizes shearing.
Specifications

Specifications

0.5 mm Yttria-stabilized Zirconium Oxide Beads
FastPrep instruments or other bead beaters
Lysing Matrix Y
Yeast, Fungi, Algea
Nucleic Acid Isolation and Purification, Homogenization
Videos
SDS
Documents

Documents

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.