missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine Hydrochloride, Granular, USP, 98.5-101.5%, Spectrum™ Chemical
L1142, 657-27-2, C6H14N2O2·HCl
Chemical Identifiers
| CAS | 657-27-2 |
|---|---|
| Molecular Formula | C6H15ClN2O2 |
| Molecular Weight (g/mol) | 182.65 |
| MDL Number | MFCD00064564 MFCD00081870 |
| InChI Key | BVHLGVCQOALMSV-JEDNCBNOSA-N |
| IUPAC Name | (2S)-2,6-diaminohexanoic acid hydrochloride |
| SMILES | Cl.NCCCC[C@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18603993
|
N/A | 100 g | Poly Bottle |
N/A
|
|
||||
|
18603994
|
N/A | 500 g | Poly Bottle |
N/A
|
|
||||
|
18603995
|
N/A | 1 kg | Poly Pail |
N/A
|
|
||||
|
18603996
|
N/A | 5 kg | Poly Pail |
N/A
|
|
||||
|
18603997
|
N/A | 50 kg | Fiber Drum |
N/A
|
|
||||
Description
Spectrum™ Chemical Lysine Hydrochloride, Granular, USP is an amino acid and protein building block. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 657-27-2 | |
| 182.65 | |
| BVHLGVCQOALMSV-JEDNCBNOSA-N | |
| Cl.NCCCC[C@H](N)C(O)=O |
| C6H15ClN2O2 | |
| MFCD00064564 MFCD00081870 | |
| (2S)-2,6-diaminohexanoic acid hydrochloride |
Specifications
| 657-27-2 | |
| 0.1% | |
| 0.4% | |
| MFCD00064564 MFCD00081870 | |
| 0.03% | |
| Cl.NCCCC[C@H](N)C(O)=O | |
| 182.65 | |
| USP |
| 100% | |
| 30 ppm | |
| C6H15ClN2O2 | |
| +20.4° to +21.4° | |
| BVHLGVCQOALMSV-JEDNCBNOSA-N | |
| (2S)-2,6-diaminohexanoic acid hydrochloride | |
| 98.5 to 101.5% |