missing translation for 'onlineSavingsMsg'
Learn More

Lysine Hydrochloride, Granular, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603994
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603994 500 g Poly Bottle
18603993 100 g Poly Bottle
18603995 1 kg Poly Pail
18603996 5 kg Poly Pail
18603997 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18603994 Supplier Spectrum Chemical Mfg Cor Supplier No. L1142500GM
Only null left

L1142, 657-27-2, C6H14N2O2·HCl

Spectrum™ Chemical Lysine Hydrochloride, Granular, USP is an amino acid and protein building block. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064564 MFCD00081870
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
IUPAC Name (2S)-2,6-diaminohexanoic acid hydrochloride
SMILES Cl.NCCCC[C@H](N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Iron (Fe) 30 ppm
Loss on Drying 0.4%
Packaging Poly Bottle
Specific Rotation +20.4° to +21.4°
Sulfate 0.03%
Quantity 500 g
Percent Purity 98.5 to 101.5%
Grade USP