missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Lyovapor™ L-200 Pro Control Freeze Dryer
The compact Lyovapor™ L-200 Pro Control offers high-quality Freeze Drying with complete automation.
$65,736.04 - $89,972.45
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|
05403005
|
BUCHI
11L200PH2422111002 |
Heatable Shelves 4, Manifold Top 12, Sensor, Software, Pfeiffer Duo 6 Vacuum Pump |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
05405435
|
BUCHI
11L200PH2412001002 |
Heatable Shelves 4, Pfeiffer Duo 6 Vacuum Pump |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
05405436
|
BUCHI
11L200PH2422113002 |
Heatable Shelves 4, Manifold Top 12, Sensor, Software, Edwards nXDS6iC Vacuum Pump |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
05405437
|
BUCHI
11L200PH2432110002 |
Heatable Shelves 4, Stoppering Top, Sensor, Software |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
05403007
|
BUCHI
11L200PH2432111002 |
Heatable Shelves 4, Stoppering Top, Sensor, Software, Pfeiffer Duo 6 Vacuum Pump |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
05405438
|
BUCHI
11L200PH2612101102 |
Heatable Shelves 6, Sensor, Pfeiffer Duo 6 Vacuum Pump, Mobile Trolley |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- A 6L condenser capacity allows for maximum sample throughput
- Integrating Infinite-Control™ with your freeze dryer gives you access to all process parameters and keeps you informed by push notifications anywhere, anytime
- Fully Automated
- Method creation and reporting
- Real-time pressure and temperature recording
- Stoppering available under inert atmospheric conditions
- Small footprint
- Touch-screen control panel
Certifications
CE Certified
Specifications
| 208/230 V |