missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ LY 2087101
Description
Allosteric potentiator of α7, α4β2 and α4β4 nAChRs; displays selectivity against α3β4 nAChRs. Thought to potentiate agonist-evoked α7 responses by binding within the nAChR transmembrane region.
Specifications
Specifications
| Quantity | 10 mg |
| Product Line | Tocris |
| Formula Weight | 318.39 |
| Purity | >98% |
| Molecular Weight (g/mol) | 318.384 |
| Chemical Name or Material | LY 2087101 |
Safety and Handling
| Recommended Storage | Store at +4°C |