missing translation for 'onlineSavingsMsg'
Learn More

Lutein, ≥96% (HPLC), MilliporeSigma™ Supelco™

Lutein is a yellow dihydroxylated carotenoid which is found to be a common constituent in many dietary supplements.

$960.50 - $3367.36

Specifications

CAS 127-40-2
Physical Form Powder
Linear Formula C40H56O2
MDL Number MFCD08435941
Synonym Xanthophyll; alpha-Carotene-3,3′-diol; beta, epsilon-carotene; beta, epsilon-carotene-3,3′-diol; Lutein
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012895
View Documents
Merck Emd Millipore
071681MG
1 mg
Each for $960.50
Only null left
 
111012896
View Documents
Merck Emd Millipore
071685MG
5 mg
Each for $3,367.36
Only null left
 
Description

Description

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

127-40-2
C40H56O2
Xanthophyll; alpha-Carotene-3,3′-diol; beta, epsilon-carotene; beta, epsilon-carotene-3,3′-diol; Lutein
568.87
Analytical Standard
Powder
MFCD08435941
Analytical Standard
≥96% (HPLC)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'