missing translation for 'onlineSavingsMsg'
Learn More

Lutein, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012896
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012896 5 mg
111012895 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012896 Supplier Merck Emd Millipore Supplier No. 071685MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lutein is a yellow dihydroxylated carotenoid which is found to be a common constituent in many dietary supplements.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 127-40-2
Physical Form Powder
Linear Formula C40H56O2
MDL Number MFCD08435941
Synonym Xanthophyll; alpha-Carotene-3,3′-diol; beta, epsilon-carotene; beta, epsilon-carotene-3,3′-diol; Lutein
Type Analytical Standard
Quantity 5 mg
Formula Weight 568.87
Percent Purity ≥96% (HPLC)
Grade Analytical Standard