Learn More
Cytiva Luer Connectors for Chromatography Columns
For use with the HiTrap column family of columns
$103.55 - $129.29
| Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
|
45-000-415
|
Cytiva
18100368 |
Tubing Connector |
Pack of 2 Each for $129.29
|
|
|||||
|
45-000-440
|
Cytiva
18102762 |
Union, Bottom |
Pack of 2 Each for $103.55
|
|
|||||
Description
- HiTrap column inlets have 1/16 in. female threads, while the outlets have 1/16 in. male threads
Specifications
| 2 Pack | |
| Polypropylene | |
| Column Accesory |