missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Low Temperature Receiving Flask

Low temperature receiving flasks are used for cold trap and other low temperature applications.

$587.19 - $1,362.19

Specifications

Color Clear
Connection Type 35/20 Joint
Material Borosilicate Glass (3.3)
Type Receiving Flask
For Use With (Application) Collecting Condensed Solvents
View More Specs

Products 4
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
05405059
View Documents Promotion Details
BUCHI
11060908
250 mL
Each for $626.72
Only null left
 
05405031
View Documents Promotion Details
BUCHI
040774
500 mL
Each for $723.59
Only null left
 
05405032
View Documents Promotion Details
BUCHI
040775
1000 mL
Each for $587.19
Only null left
 
05405033
View Documents Promotion Details
BUCHI
040777
3000 mL
Each for $1,362.19
Only null left
 
Description

Description

  • Spherical flasks with ball joint (35/20) for collecting condensed solvents
  • Plastic+Glas (P+G) coating for maximized safety
  • High chemical resistance against acids, alkalis, and organic substances
  • Operating range: -70° to 40°C
Specifications

Specifications

Clear
Borosilicate Glass (3.3)
Collecting Condensed Solvents
35/20 Joint
Receiving Flask
70°C to 40°C
Videos
SDS
Documents

Documents