missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Lovastatin | |
| 179°C | |
| C24H36O5 | |
| 25 g | |
| PCZOHLXUXFIOCF-BXMDZJJMSA-N | |
| (1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate | |
| 53232 | |
| 404.55 | |
| Crystalline Powder |
| 75330-75-5 | |
| White-Yellow | |
| MFCD00072164 | |
| lovastatin, mevinolin, mevacor, monacolin k, lovalip, altoprev, lovalord, mevinacor, nergadan, 6alpha-methylcompactin | |
| CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12 | |
| 404.55 | |
| CHEBI:40303 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 75330-75-5 | |
| 404.55 | |
| PCZOHLXUXFIOCF-BXMDZJJMSA-N | |
| 53232 | |
| (1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate |
| C24H36O5 | |
| MFCD00072164 | |
| lovastatin, mevinolin, mevacor, monacolin k, lovalip, altoprev, lovalord, mevinacor, nergadan, 6alpha-methylcompactin | |
| CHEBI:40303 | |
| CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12 |
Safety and Handling
RTECSNumber : EK7907000
TSCA : No