missing translation for 'onlineSavingsMsg'
Learn More

Lovastatin 98.0+%, TCI America™

Catalog No. p-7131870
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02145G 5 g
L021425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02145G Supplier TCI America Supplier No. L02145G
Only null left

Chemical Identifiers

CAS 75330-75-5
Molecular Formula C24H36O5
Molecular Weight (g/mol) 404.55
MDL Number MFCD00072164
InChI Key PCZOHLXUXFIOCF-BXMDZJJMSA-N
Synonym lovastatin, mevinolin, mevacor, monacolin k, lovalip, altoprev, lovalord, mevinacor, nergadan, 6alpha-methylcompactin
PubChem CID 53232
ChEBI CHEBI:40303
IUPAC Name (1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 5 g
Formula Weight 404.55
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Lovastatin
RTECSNumber EK7907000
TSCA No