accessibility menu, dialog, popup

UserName

CAS RN 75330-75-5

IUPAC Name: (1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate
Synonyms: lovastatin lovastatin preparation lipofren lovastatin, (1 α(s*))-isomer colevix lovastatine lovastatina 6 methylcompactin nergadan (1 α(S*))-isomer lovastatin
Molecular Weight (g/mol): 404.55
Molecular Formula: C24H36O5
InChi Key: PCZOHLXUXFIOCF-BXMDZJJMSA-N
SMILES: CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)