missing translation for 'onlineSavingsMsg'
Learn More

Loperamide Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America L015425G

 View more versions of this product

Catalog No. L015425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Loperamide Hydrochloride
223°C
C29H34Cl2N2O2
25 g
loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec
CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl
513.503
CHEBI:6533
≥98.0% (T,HPLC)
34552-83-5
White
MFCD00058581
2811
PGYPOBZJRVSMDS-UHFFFAOYSA-N
4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
71420
513.50
Crystalline Powder

Chemical Identifiers

34552-83-5
513.503
PGYPOBZJRVSMDS-UHFFFAOYSA-N
71420
4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
C29H34Cl2N2O2
MFCD00058581
loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec
CHEBI:6533
CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TM4960000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions