missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Lomustine | |
| 91°C | |
| C9H16ClN3O2 | |
| 200 mg | |
| 1-(2-Chloroethyl)-3-cyclohexyl-1-nitrosourea | |
| ClCCN(N=O)C(=O)NC1CCCCC1 | |
| 233.70 | |
| CHEBI:6520 | |
| ≥98.0% (HPLC) |
| 13010-47-4 | |
| Yellow | |
| MFCD00012392 | |
| 2811 | |
| GQYIWUVLTXOXAJ-UHFFFAOYSA-N | |
| 3-(2-chloroethyl)-1-cyclohexyl-3-nitrosourea | |
| 3950 | |
| 233.70 | |
| Crystalline Powder |
Chemical Identifiers
| 13010-47-4 | |
| 233.70 | |
| GQYIWUVLTXOXAJ-UHFFFAOYSA-N | |
| 3950 | |
| 3-(2-chloroethyl)-1-cyclohexyl-3-nitrosourea |
| C9H16ClN3O2 | |
| MFCD00012392 | |
| 1-(2-Chloroethyl)-3-cyclohexyl-1-nitrosourea | |
| CHEBI:6520 | |
| ClCCN(N=O)C(=O)NC1CCCCC1 |
Safety and Handling
RTECSNumber : YS4900000
TSCA : No
Recommended Storage : Freezer