missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lomustine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 91°C |
| Color | Yellow |
| Quantity | 50 mg |
| UN Number | 2811 |
| Formula Weight | 233.70 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Lomustine |
Safety and Handling
| RTECSNumber | YS4900000 |
| TSCA | No |
| Recommended Storage | Freezer |