accessibility menu, dialog, popup

UserName

CAS RN 13010-47-4

ClNNOOHN

CAS RN 13010-47-4

IUPAC Name: 3-(2-chloroethyl)-1-cyclohexyl-3-nitrosourea
Synonyms: lomustine lomustina [inn-spanish] lomustine preparation lomustine (product) belustine bristol myers squibb brand of lomustine C9H16clN3O2 cecenu lomustine (substance) 洛莫司汀
Molecular Weight (g/mol): 233.7
Molecular Formula: C9H16ClN3O2
InChi Key: GQYIWUVLTXOXAJ-UHFFFAOYSA-N
SMILES: ClCCN(N=O)C(=O)NC1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results