missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lomerizine Dihydrochloride 98.0+%, TCI America™
Supplier: TCI America L02451G
Specifications
| Lomerizine Dihydrochloride | |
| 180°C | |
| C27H32Cl2F2N2O3 | |
| 1 g | |
| LOGVKVSFYBBUAJ-UHFFFAOYSA-N | |
| dihydrogen 1-[bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine dichloride | |
| 122125 | |
| ≥98.0% (HPLC,T) |
| 101477-54-7 | |
| White | |
| MFCD01703867 | |
| 1-[Bis(4-fluorophenyl)methyl]-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride | |
| [H+].[H+].[Cl-].[Cl-].COC1=CC=C(CN2CCN(CC2)C(C2=CC=C(F)C=C2)C2=CC=C(F)C=C2)C(OC)=C1OC | |
| 541.46 | |
| 541.46 | |
| Crystalline Powder |
Chemical Identifiers
| 101477-54-7 | |
| 541.46 | |
| LOGVKVSFYBBUAJ-UHFFFAOYSA-N | |
| 122125 | |
| [H+].[H+].[Cl-].[Cl-].COC1=CC=C(CN2CCN(CC2)C(C2=CC=C(F)C=C2)C2=CC=C(F)C=C2)C(OC)=C1OC |
| C27H32Cl2F2N2O3 | |
| MFCD01703867 | |
| 1-[Bis(4-fluorophenyl)methyl]-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride | |
| dihydrogen 1-[bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine dichloride |
Safety and Handling
RTECSNumber : TK9189000
TSCA : No
Recommended Storage : Refrigerator