missing translation for 'onlineSavingsMsg'
Learn More

Lomerizine Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7131890
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0245200MG 200 mg
L02451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0245200MG Supplier TCI America Supplier No. L0245200MG
Only null left

Chemical Identifiers

CAS 101477-54-7
Molecular Formula C27H32Cl2F2N2O3
Molecular Weight (g/mol) 541.46
MDL Number MFCD01703867
InChI Key LOGVKVSFYBBUAJ-UHFFFAOYSA-N
Synonym 1-[Bis(4-fluorophenyl)methyl]-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride
PubChem CID 122125
IUPAC Name dihydrogen 1-[bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].COC1=CC=C(CN2CCN(CC2)C(C2=CC=C(F)C=C2)C2=CC=C(F)C=C2)C(OC)=C1OC

Specifications

Melting Point 180°C
Color White
Quantity 200 mg
Formula Weight 541.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Lomerizine Dihydrochloride
RTECSNumber TK9189000
TSCA No
Recommended Storage Refrigerator