missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Locust bean gum

Catalog No. AC466670010
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466670010 1 kg
AC466672500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466670010 Supplier Thermo Scientific Chemicals Supplier No. 466670010

CAS: 9000-40-2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9000-40-2

Specifications

Quantity 1 kg
Chemical Name or Material Locust bean gum
EINECSNumber 232-541-5

RUO – Research Use Only