missing translation for 'onlineSavingsMsg'
Learn More

Lithium Salicylate Monohydrate 98.0+%, TCI America™

Catalog No. L028525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L028525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L028525G Supplier TCI America Supplier No. L028525G
Only null left

Chemical Identifiers

CAS 75575-50-7
Molecular Formula C7H7LiO4
Molecular Weight (g/mol) 162.069
InChI Key SKQWWJVTSPXPSF-UHFFFAOYSA-M
Synonym Salicylic Acid Lithium Salt
PubChem CID 131674850
IUPAC Name lithium;2-carboxyphenolate;hydrate
SMILES [Li+].C1=CC=C(C(=C1)C(=O)O)[O-].O

Specifications

Quantity 25 g
Formula Weight 144.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium Salicylate Monohydrate