accessibility menu, dialog, popup

UserName

LiOHOOH2O

CAS RN 75575-50-7

IUPAC Name: lithium(1+) 2-carboxybenzen-1-olate hydrate
Synonyms: lithium(1+) 2-carboxybenzenolate hydrate
Molecular Weight (g/mol): 162.07
Molecular Formula: C7H7LiO4
InChi Key: SKQWWJVTSPXPSF-UHFFFAOYSA-M
SMILES: [Li+].O.OC(=O)C1=CC=CC=C1[O-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results