missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide, Monohydrate, Crystal, ACS, 98%, Spectrum™ Chemical

$337.97 - $15170.13

Chemical Identifiers

CAS 1310-66-3
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611320
View Documents
Spectrum Chemical Mfg Cor
L1156125GM
125 g Amber Glass Bottle
Each for $337.97
Only null left
 
18611321
View Documents
Spectrum Chemical Mfg Cor
L1156500GM
500 g Amber Glass Bottle
Each for $782.00
Only null left
 
18611322
View Documents
Spectrum Chemical Mfg Cor
L115625KG
2.5 kg Amber Glass Bottle
Each for $1,896.53
Only null left
 
18611323
View Documents
Spectrum Chemical Mfg Cor
L115612KG
12 kg Poly Pail
Each for $6,443.00
Only null left
 
18611324
View Documents
Spectrum Chemical Mfg Cor
L115650KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1310-66-3

Specifications

1310-66-3
125 g
ACS
1
98%
Amber Glass Bottle
Videos
SDS
Documents

Documents

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.