missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Hydroxide, Monohydrate, Crystal, ACS, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 50 kg |
| Percent Purity | 98% |
| Grade | ACS |
| Packaging | Fiber Drum |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.