missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide, Monohydrate, Crystal, ACS, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L115625KG

 View more versions of this product

Catalog No. 18611322


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1310-66-3
2.5 kg
ACS
1
98%
Amber Glass Bottle

Chemical Identifiers

1310-66-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.