missing translation for 'onlineSavingsMsg'
Learn More

Liranaftate 98.0+%, TCI America™

Catalog No. L030150MG
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L030150MG 50 mg
L0301250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L030150MG Supplier TCI America Supplier No. L030150MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88678-31-3
Molecular Formula C18H20N2O2S
Molecular Weight (g/mol) 328.43
MDL Number MFCD00865576
InChI Key VPHPQNGOVQYUMG-UHFFFAOYSA-N
Synonym O-(5,6,7,8-Tetrahydronaphthalen-2-yl) (6-Methoxypyridin-2-yl)(methyl)carbamothioate, (6-Methoxypyridin-2-yl)(methyl)carbamothioic Acid O-(5,6,7,8-Tetrahydronaphthalen-2-yl) Ester
PubChem CID 3936
IUPAC Name N-(6-methoxypyridin-2-yl)-N-methyl-1-(5,6,7,8-tetrahydronaphthalen-2-yloxy)methanethioamide
SMILES COC1=CC=CC(=N1)N(C)C(=S)OC1=CC=C2CCCCC2=C1

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 50 mg
Formula Weight 328.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Liranaftate
RTECSNumber FD3890000