accessibility menu, dialog, popup

UserName

CAS RN 88678-31-3

IUPAC Name: N-(6-methoxypyridin-2-yl)-N-methyl-1-(5,6,7,8-tetrahydronaphthalen-2-yloxy)methanethioamide
Synonyms: liranaftate 利拉萘酯 liranaftato piritetrate zefnart liranaftatum
Molecular Weight (g/mol): 328.43
Molecular Formula: C18H20N2O2S
InChi Key: VPHPQNGOVQYUMG-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=N1)N(C)C(=S)OC1=CC=C2CCCCC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)