Learn More
MilliporeSigma™ Supelco™ Liquid Scintillation Vials
Polyethylene with light-weight walls for increased counting efficiency.
$471.46 - $838.86
Specifications
Product Type Screw Thread Vial
View More Specs
Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
111009032
View Documents Promotion Details
MilliporeSigma™ Supelco™
V67551000EA
Cap size 24 mm
Each for $838.86
Only null left
111009829
View Documents Promotion Details
MilliporeSigma™ Supelco™
V6880500EA
Cap size 22 mm
Each for $471.46
Only null left
Description
Description
  • Polyethylene with light-weight walls for increased counting efficiency.
  • Vials and caps are packed in separate plastic bags.
Specifications
Specifications
Screw Thread Vial