missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liquefied Phenol, USP, 89%, Spectrum™ Chemical
PH125, 108-95-2, C6H6O
Supplier: Spectrum Chemical Mfg Cor PH1254LTPVCS4
Description
Spectrum™ Chemical Liquefied Phenol, USP is commonly used for permanent treatment of ingrown toe and finger nails, a procedure known as a chemical matrixectomy. All Spectrum Chemical USP products USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Spécifications
| 108-95-2,7732-18-5 | |
| Poly-Coated Glass Bottle | |
| MFCD00002143 | |
| UN2821 | |
| OC1=CC=CC=C1 | |
| 94.11 | |
| USP |
| 89%,11% | |
| C6H6O | |
| 4 x 4 L | |
| ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
| phenol | |
| 89% | |
| Liquid |
Identifiants chimiques
| 108-95-2 | |
| 94.11 | |
| ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
| OC1=CC=CC=C1 |
| C6H6O | |
| MFCD00002143 | |
| phenol |