missing translation for 'onlineSavingsMsg'
Learn More

Liquefied Phenol, USP, 89%, Spectrum™ Chemical

PH125, 108-95-2, C6H6O

Supplier:  Spectrum Chemical Mfg Cor PH1254LTPV

 View more versions of this product

Catalog No. 18603908

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Liquefied Phenol, USP is commonly used for permanent treatment of ingrown toe and finger nails, a procedure known as a chemical matrixectomy. All Spectrum Chemical USP products USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

108-95-2,7732-18-5
Poly-Coated Glass Bottle
MFCD00002143
UN2821
OC1=CC=CC=C1
94.11
USP
89%,11%
C6H6O
4 L
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
89%
Liquid

Chemical Identifiers

108-95-2
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
OC1=CC=CC=C1
C6H6O
MFCD00002143
phenol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions