missing translation for 'onlineSavingsMsg'
Learn More

Lipid Removal Agent (LRA), MilliporeSigma™ Supelco™

LRA, the synthetic adsorbent of crystalline calcium silicate hydrate is used for removing lipids and endotoxin from plasma or aqueous solutions. It is also used for DNA purification to remove protein, RNA and genomic DNA.

$416.17 - $1311.79

Specifications

Grade Reagent Grade
Physical Form Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111020854
View Documents
Merck Emd Millipore
13358U
100 g
Each for $416.17
Only null left
 
111003659
View Documents
Merck Emd Millipore
13360U
500 g
Each for $1,311.79
Only null left
 
Description

Description

Specifications

Specifications

Reagent Grade
Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'