Learn More
Lipid Removal Agent (LRA), MilliporeSigma™ Supelco™
LRA, the synthetic adsorbent of crystalline calcium silicate hydrate is used for removing lipids and endotoxin from plasma or aqueous solutions. It is also used for DNA purification to remove protein, RNA and genomic DNA.
Supplier: Merck Emd Millipore 13360U
Specifications
| 500 g | |
| Reagent Grade | |
| Powder |