missing translation for 'onlineSavingsMsg'
Learn More

Lipid Removal Agent (LRA), MilliporeSigma™ Supelco™

Catalog No. 111020854
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020854 100 g
111003659 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020854 Supplier Merck Emd Millipore Supplier No. 13358U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

LRA, the synthetic adsorbent of crystalline calcium silicate hydrate is used for removing lipids and endotoxin from plasma or aqueous solutions. It is also used for DNA purification to remove protein, RNA and genomic DNA.

Specifications

Quantity 100 g
Grade Reagent Grade
Physical Form Powder