missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linoleic acid, ≥98% (GC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 6223025MLF
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal CareSpecifications
| 60-33-3 | |
| >113°C - closed cup | |
| CH3(CH2)4 CH=CHCH2 CH=CH(CH2)7 CO2 H | |
| -5°C | |
| 9-cis,12-cis-Linoleic acid; cis-9,cis-12-Octadecadienoic acid; Telfairic acid | |
| 25 mL | |
| ≥98% (GC) |
| 0.902 g/mL (at 25°C) | |
| Neat | |
| MFCD00064241 | |
| 229°C to 230°C (16 mmHg) | |
| Analytical Standard | |
| 280.45 | |
| Analytical Standard |