missing translation for 'onlineSavingsMsg'
Learn More

Linoleic acid, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187595
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014856 5 mL
111014855 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014856 Supplier Merck Emd Millipore Supplier No. 622305MLF
Only null left
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 60-33-3
Density 0.902 g/mL (at 25°C)
Flash Point >113°C - closed cup
Physical Form Neat
Linear Formula CH3(CH2)4 CH=CHCH2 CH=CH(CH2)7 CO2 H
MDL Number MFCD00064241
Melting Point -5°C
Boiling Point 229°C to 230°C (16 mmHg)
Synonym 9-cis,12-cis-Linoleic acid; cis-9,cis-12-Octadecadienoic acid; Telfairic acid
Type Analytical Standard
Quantity 5 mL
Formula Weight 280.45
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less