missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Linear polyacrylamide solution for DNA/RNA precipitation, 5mg/ml, Molecular Biology Grade
Supplier: Thermo Scientific Chemicals J67830XF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Clear, colorless | |
| Linear polyacrylamide solution | |
| LPA solution for DNA/RNA precipitation, Molecular Biology Grade; |
| -30°C to -10°C | |
| 1 mL | |
| Liquid |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only