missing translation for 'onlineSavingsMsg'
Learn More

Linear Alkylbenzene Sulfonate Standard, 1000 ppm LAS, Ricca Chemical

$910.33 - $1672.13

Chemical Identifiers

CAS 68411-30-3
Molecular Formula C6H5NaO3S
Molecular Weight (g/mol) 180.15
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
435016
View Documents
Ricca Chemical Company
435016
500 mL Amber Glass Bottle
Each for $910.33
Only null left
 
435032
View Documents
Ricca Chemical Company
435032
1 L Amber Glass Bottle
Each for $1,672.13
Only null left
 
Description

Description

Specifications

Chemical Identifiers

68411-30-3
C6H5NaO3S
180.15

Specifications

Amber Glass Bottle
98.0,0.1,0.08
100.0,0.1,0.08
Liquid
Colorless
Standard
500 mL
68411-30-3,7664-93-9,68411-30-3,7732-18-5
1,000ppm LAS
1 mL = 1 mg LAS
C6H5NaO3S
Approximately 0°C
180.15
Linear Alkylbenzene Sulfonate (LAS) Standard
Videos
SDS
Documents

Documents