missing translation for 'onlineSavingsMsg'
Learn More

Linear Alkylbenzene Sulfonate Standard, 1000 ppm LAS, Ricca Chemical

Catalog No. 435016
missing translation for 'changeView'
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
435016 500 mL Amber Glass Bottle
435032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 435016 Supplier Ricca Chemical Company missing translation for 'supplierNo' 435016
Only null left

Chemical Identifiers

CAS 68411-30-3 , 7664-93-9 , 68411-30-3 , 7732-18-5
Molecular Formula C6H5NaO3S
Molecular Weight (g/mol) 180.15

Specifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 1,000ppm LAS
CAS Min % 98.0, 0.1, 0.08
CAS Max % 100.0, 0.1, 0.08
Equivalence 1 mL = 1 mg LAS
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Chemical Name or Material Linear Alkylbenzene Sulfonate (LAS) Standard
Show More Show Less