missing translation for 'onlineSavingsMsg'
Learn More

Linear Alkylbenzene Sulfonate Standard, 1000 ppm LAS, Ricca Chemical

Catalog No. 43504
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43504 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43504 Supplier Ricca Chemical Company Supplier No. 43504

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9 , 68411-30-3
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

CAS Min % 98.0, 0.1, 0.08
CAS Max % 100.0, 0.1, 0.08
Equivalence 1 mL = 1 mg LAS
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1000 ppm LAS
Packaging Amber Glass Bottle
pH 1.5 to 2
Quantity 120 mL
Chemical Name or Material Linear Alkybenzene Sulfonate (LAS) Solution