missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Linbro™ 2.0mL Snap-Top Microcentrifuge Tubes

With reference lines at 0.5, 1.0, 1.5 and 2.0 mL for quick volume estimations

Supplier:  MP Biomedicals™ 0219352990

 View more versions of this product

Catalog No. ICN19352990


Only null left
Explore available promotions

Description

Description

Frosted writing area.

Specifications

Specifications

Snap Cap
Natural
For those experiments that entail large numbers of samples that may be difficult to organize and keep track of
Snap Cap
2 mL
Plastic
500/Cs.
0.5, 1, 1.5 and 2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.