missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Light Emission Accelerator-II
Description
Light emission triggering solution with luminescence enhancer for use with the Galacto-Light Plus™ or Dual-Light™ Reporter Gene Assay Systems.
Light emission triggering solution with luminescence enhancer for use with the Galacto-Light Plus™ or Dual-Light™ Reporter Gene Assay Systems.
For Research Use Only. Not for use in diagnostics procedures.
- Form: Liquid
- Format: Bottle
- Substrate: Alkaline Phosphatase
Specifications
Specifications
| Product Type | β-Galactosidase Reporter Gene Assay System |
| Content And Storage | Ambient for shipping. Store the product at 4°C. |
| Shipping Condition | Room Temperature |
| Substrate | Alkaline Phosphatase |
| Substrate Type | AP (Alkaline Phosphatase) Substrate |
| Product Line | NovaBright |
| Quantity | 210 mL |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.