missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Life Sciences Protein G GraviTrap™
Description
Prepacked gravity-flow columns provide convenience, save time, and increase the reliability of your results.
- Purification of antibodies in 30 min allows you to quickly obtain and store your target protein in order to avoid the inherent dangers of protein instability and degradation associated with slow and long purification runs
- High purity and recovery with purity > 95% in one step and recovery typically 70% to 80%
- High binding capacity such as Protein G GraviTrap approximately 20mg human IgG/column
- Complete pack size: 10 columns
- Application: Manual purification of monoclonal and polyclonal antibodies
- Includes: 1 pack of 10 × 1mL columns
- Binding capacity: Approx. 20mg human IgG/column
- Bed volume: 1mL
- Material (Column hardware): Polypropylene barrel, Polyethylene frits
Specifications
Specifications
| Bed Volume | 1 mL |
| Description | Protein G GraviTrap includes ten columns with 1 mL of resin for general purpose purification of polyclonal and monoclonal antibodies from most species. |
| Kit Contents | 10 Ready-to -use column prepacked with 1mL of Protein G Sepharose 4 fast flow, Polypropylene barrel, Polyethylene frits |
| Content And Storage | 2°C to 8°C |
| Quantity | 10 Pack |
| For Use With (Application) | Fast and efficient manual purification of monoclonal and polyclonal antibodies |
| Type | Gravity Flow Column |
| Capacity (Metric) | 1 μL |