missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium 1-Hexanesulfonate monohydrate , ≥99.0% (T), MilliporeSigma™ Supelco™

$295.78 - $1143.59

Chemical Identifiers

MDL Number MFCD00149549
Synonym 1-Hexanesulfonic acid sodium salt monohydrate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111013499
View Documents
Merck Emd Millipore
5286210GF
10 g
Each for $295.78
Only null left
 
111013500
View Documents
Merck Emd Millipore
5286250GF
50 g
Each for $1,143.59
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00149549
1-Hexanesulfonic acid sodium salt monohydrate

Specifications

207300-91-2
MFCD00149549
1-Hexanesulfonic acid sodium salt monohydrate
≥99.0% (T)
LiChropur
CH3(CH2)4 CH2 SO3 Na · H2O
10 g
206.24
For ion pair chromatography
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany