missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Potassium Phosphate Monobasic, MilliporeSigma™ Supelco™

Catalog No. 111022678
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022678 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022678 Supplier Merck Emd Millipore Supplier No. 57618250G

For HPLC

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym Monopotassium phosphate; Potassium dihydrogen phosphate; prim.-Potassium phosphate
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 252.6°C (literature)
Density 2.338 g/mL (at 25°C (literature))
pH 4.1 to 4.5
Quantity 250 g
Linear Formula KH2PO4
UN Number NONH for all modes of transport
Solubility Information H2O: 10%, Clear, Colorless
Formula Weight 136.09
Percent Purity 99.5 to 101% (T)
Grade HPLC
RTECSNumber TC6615500

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany