missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Hexadecyltrimethylammonium bromide, ≥99.0% (AT), MilliporeSigma™ Supelco™
Hexadecyltrimethylammonium bromide (CTAB) is a quaternary ammonium salt containing one long hydrophobic alkyl chain called amphiphile. The hydrophobic ions interact forming discrete aggregates possessing a hydrophobic core and a polar surface, called micelles in polar solution.
$331.91 - $1288.67
Chemical Identifiers
| MDL Number | MFCD00011772 |
|---|---|
| Synonym | CTAB; Cetrimonium bromide; Cetyltrimethylammonium bromide; Palmityltrimethylammonium bromide |
Chemical Identifiers
| MFCD00011772 | |
| CTAB; Cetrimonium bromide; Cetyltrimethylammonium bromide; Palmityltrimethylammonium bromide |
Specifications
| 248°C to 251°C | |
| Powder or Crystals | |
| CH3(CH2)15 N(Br)(CH3)3 | |
| 244°C | |
| Water: soluble 36.4 g/L (at 20°C (completely)) | |
| ≥99.0% (AT) | |
| LiChropur |
| 57-09-0 | |
| 10 g | |
| MFCD00011772 | |
| CTAB; Cetrimonium bromide; Cetyltrimethylammonium bromide; Palmityltrimethylammonium bromide | |
| 364.45 | |
| Ion Pair Chromatography |
Safety and Handling
RTECSNumber : BQ7875000
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany