Learn More
Thermo Scientific™ LH-4000W 8 x 500 mL Windshielded Swinging Bucket Rotor
Support a diverse array of sample processing applications without the need to modify existing protocols.
$416.34 - $41,372.64
Specifications
For Use With (Equipment) Thermo Scientific Sorvall RC 4 Series, Cryofuge 5500i and Multifuge 4 Large Capacity Centrifuges
Biocontainment Yes
View More Specs
Products 4
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
75006421
View Documents Promotion Details
Thermo Scientific™
75006421
Sealing Cap
Pack of 2 Each for $1,969.02
Only null left
50868601
View Documents Promotion Details
Thermo Scientific™
75006476
Swinging Bucket Rotor N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
50868615
View Documents Promotion Details
Thermo Scientific™
75006477
Bucket Rotor
Each for $1,965.26
Only null left
50868614
View Documents Promotion Details
Thermo Scientific™
75006486
Microplate Adapter
Each for $416.34
Only null left
Description
Description
Support a diverse array of sample processing applications without the need to modify existing protocols with the Thermo Scientific™ LH-4000W 8 x 500mL Windshield Swinging Bucket Rotor. Choose from round, DoubleSpin™ and Double blood bag buckets with a wide array of available adapters, providing capacity flexibility ranging from 1.5 to 250mL.
Ordering Information:
Buckets, biocontainment lids and DoubleSpin buckets are sold separately; order rotor body (75006476) and components to complete assembly. Tube adapters and microplate adapter for DoubleSpin bucket are also available separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific™ Cryofuge 5500i series centrifuges
Specifications
Specifications
Thermo Scientific Sorvall RC 4 Series, Cryofuge 5500i and Multifuge 4 Large Capacity Centrifuges
Yes