missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Levocetirizine dihydrochloride

$107.87 - $345.80

Chemical Identifiers

CAS 130018-87-0
Molecular Formula C21H25ClN2O3·2HCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC467470010
View Documents
Thermo Scientific Chemicals
467470010
1 g
Each for $107.87
Only null left
 
AC467470050
View Documents
Thermo Scientific Chemicals
467470050
5 g
Each for $345.80
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

130018-87-0
C21H25ClN2O3·2HCl

Specifications

130018-87-0
Solubility aq. soln.: soluble.
C21H25ClN2O3·2HCl
PGLIUCLTXOYQMV-UHFFFAOYNA-N
2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dihydrochloride
Levocetirizine dihydrochloride
1 g
214.0°C
461.81
Cl.Cl.OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
461.81
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

Very toxic to aquatic life.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Avoid release to the environment.

Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only