missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Levamisole hydrochloride, 99+%

CAS: 16595-80-5 | C11H12N2S·ClH | 240.76 g/mol

$149.03 - $149.03

Chemical Identifiers

CAS 16595-80-5
Molecular Formula C11H12N2S·ClH
Molecular Weight (g/mol) 240.76
MDL Number MFCD00012675
InChI Key LAZPBGZRMVRFKY-HNCPQSOCSA-N
Synonym levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris
PubChem CID 27944
ChEBI CHEBI:6433
IUPAC Name (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride
SMILES C1CSC2=NC(CN21)C3=CC=CC=C3.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC187870100
SDS View Documents
Thermo Scientific Chemicals
187870100
10 g Glass bottle
Each for $149.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16595-80-5
240.76
LAZPBGZRMVRFKY-HNCPQSOCSA-N
27944
(6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride
C11H12N2S·ClH
MFCD00012675
levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris
CHEBI:6433
C1CSC2=NC(CN21)C3=CC=CC=C3.Cl

Specifications

16595-80-5
White
Authentic
Glass bottle
MFCD00012675
15, 5511
-127.5
Solubility in water: 210g/L (20°C). Other solubilities: soluble in methanol and propylene glycol,sparingly soluble in ethanol,slightly soluble in chloroform,hexane and acetone
C1CSC2=NC(CN21)C3=CC=CC=C3.Cl
240.76
CHEBI:6433
99+%
Levamisole hydrochloride
226°C to 231°C
0.5% max. (1 g, 105°C, 4 hrs)
99+%
C11H12N2S·ClH
10 g
− 127.50 (20.00°C c=5,H2O)
levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris
LAZPBGZRMVRFKY-HNCPQSOCSA-N
(6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride
27944
240.76
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 240-654-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only