Learn More
Thermo Scientific Chemicals Levamisole hydrochloride, 99+%
CAS: 16595-80-5 | C11H12N2S·ClH | 240.76 g/mol
Supplier: Thermo Scientific Chemicals 187870100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Levamisole hydrochloride | |
| 226°C to 231°C | |
| 0.5% max. (1 g, 105°C, 4 hrs) | |
| 99+% | |
| C11H12N2S·ClH | |
| 10 g | |
| − 127.50 (20.00°C c=5,H2O) | |
| levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris | |
| LAZPBGZRMVRFKY-HNCPQSOCSA-N | |
| (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride | |
| 27944 | |
| 240.76 | |
| Crystalline Powder |
| 16595-80-5 | |
| White | |
| Authentic | |
| Glass bottle | |
| MFCD00012675 | |
| 15, 5511 | |
| -127.5 | |
| Solubility in water: 210g/L (20°C). Other solubilities: soluble in methanol and propylene glycol,sparingly soluble in ethanol,slightly soluble in chloroform,hexane and acetone | |
| C1CSC2=NC(CN21)C3=CC=CC=C3.Cl | |
| 240.76 | |
| CHEBI:6433 | |
| 99+% |
Chemical Identifiers
| 16595-80-5 | |
| 240.76 | |
| LAZPBGZRMVRFKY-HNCPQSOCSA-N | |
| 27944 | |
| (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride |
| C11H12N2S·ClH | |
| MFCD00012675 | |
| levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris | |
| CHEBI:6433 | |
| C1CSC2=NC(CN21)C3=CC=CC=C3.Cl |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 240-654-6
RUO – Research Use Only