missing translation for 'onlineSavingsMsg'
Learn More

Lercanidipine Hydrochloride 98.0+%, TCI America™

Catalog No. L0282200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0282200MG 200 mg
L02821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0282200MG Supplier TCI America Supplier No. L0282200MG
Only null left

Chemical Identifiers

CAS 132866-11-6
Molecular Formula C36H42ClN3O6
Molecular Weight (g/mol) 648.197
MDL Number MFCD07773089
InChI Key WMFYOYKPJLRMJI-UHFFFAOYSA-N
Synonym lercanidipine hydrochloride, lercanidipine hcl, cardiovasc, carmen, corifeo, lerzam, lercadip, lercapin, lercaton, lerkamen
PubChem CID 157917
IUPAC Name 5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
SMILES CC1=C(C(C(=C(N1)C)C(=O)OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl

Specifications

Melting Point 195°C
Color Yellow
Quantity 200 mg
Formula Weight 648.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Lercanidipine Hydrochloride
TSCA No
Recommended Storage Refrigerator